4-Chlorothiophenol, 98%
4-Chlorothiophenol, 98%
4-Chlorothiophenol, 98%
4-Chlorothiophenol, 98%
Thermo Scientific Chemicals
4-Chlorothiophenol, 98%
CAS: 106-54-7 | C6H5ClS | 144.616 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA154761000
Price (MYR)
440.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
440.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Chlorothiophenol
Name Note98%
CAS106-54-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger