1-Chlorobutane, 99+%, pure
1-Chlorobutane, 99+%, pure
1-Chlorobutane, 99+%, pure
1-Chlorobutane, 99+%, pure
Thermo Scientific Chemicals
1-Chlorobutane, 99+%, pure
CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA154630025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Chlorobutane
CAS109-69-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0