2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
2-Chlorobenzaldehyde, 99%
Thermo Scientific Chemicals
2-Chlorobenzaldehyde, 99%
CAS: 89-98-5 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA154615000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Chlorobenzaldehyde
Name Note99%
CAS89-98-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger