Glycolic acid, 99%
Glycolic acid, 99%
Glycolic acid, 99%
Thermo Scientific Chemicals
Glycolic acid, 99%
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA154515000
Price (MYR)
1,539.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,539.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycolic acid
CAS79-14-1
Melting Point72.0°C to 80.0°C
Boiling Point100.0°C
Health Hazard 1Danger