2-Butoxyethanol, 99%, extra pure
2-Butoxyethanol, 99%, extra pure
2-Butoxyethanol, 99%, extra pure
2-Butoxyethanol, 99%, extra pure
Thermo Scientific Chemicals
2-Butoxyethanol, 99%, extra pure
CAS: 111-76-2 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
10 L
Packaging:
View all
Metal drum
Catalog number FSA154330250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Specifications
Chemical Name or Material2-Butoxyethanol
CAS111-76-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0