Bromoethane, 98%, pure
Bromoethane, 98%, pure
Bromoethane, 98%, pure
Bromoethane, 98%, pure
Thermo Scientific Chemicals
Bromoethane, 98%, pure
CAS: 74-96-4 | C2H5Br | 108.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA154215000
Price (MYR)
376.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
376.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBromoethane
Name NotePure
CAS74-96-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
View more
Alkylation reagent
RUO – Research Use Only