2-Heptanone, 98%
2-Heptanone, 98%
2-Heptanone, 98%
Thermo Scientific Chemicals
2-Heptanone, 98%
CAS: 110-43-0 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic drum
Catalog number FSA154000051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or Material2-Heptanone
CAS110-43-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0