Anisole, 99%, pure
Anisole, 99%, pure
Anisole, 99%, pure
Thermo Scientific Chemicals
Anisole, 99%, pure
CAS: 100-66-3 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA153922500
Price (MYR)
574.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
574.00
EA
Add to cart
Specifications
Chemical Name or MaterialAnisole
CAS100-66-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven