2-Aminothiophenol, 98%
2-Aminothiophenol, 98%
2-Aminothiophenol, 98%
2-Aminothiophenol, 98%
Thermo Scientific Chemicals
2-Aminothiophenol, 98%
CAS: 137-07-5 | C6H7NS | 125.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA153901000
Price (MYR)
731.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
731.00
EA
Add to cart
Specifications
Chemical Name or Material2-Aminothiophenol
Name Note98%
CAS137-07-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger