3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure
Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure
3-Aminophthalhydrazide, 98%, pure, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0 | CAS: 521-31-3 | C8H7N3O2 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA153850250
Price (MYR)
1,278.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,278.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Aminophthalhydrazide
Name Note98%
CAS521-31-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning