4-Aminohippuric acid, 99%
4-Aminohippuric acid, 99%
4-Aminohippuric acid, 99%
4-Aminohippuric acid, 99%
Thermo Scientific Chemicals
4-Aminohippuric acid, 99%
CAS: 61-78-9 | C9H10N2O3 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA153805000
Price (MYR)
9,641.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
9,641.00
EA
Add to cart
Specifications
Chemical Name or Material4-Aminohippuric acid
CAS61-78-9
CAS Min %98.5
Health Hazard 1Warning
CAS Max %100.0