2-Aminoethanethiol hydrochloride, 98%
2-Aminoethanethiol hydrochloride, 98%
2-Aminoethanethiol hydrochloride, 98%
2-Aminoethanethiol hydrochloride, 98%
Thermo Scientific Chemicals
2-Aminoethanethiol hydrochloride, 98%
CAS: 156-57-0 | C2H8ClNS | 113.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA153771000
Price (MYR)
1,150.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,150.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Aminoethanethiol hydrochloride
CAS156-57-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0