Alizarin, 97%, pure
Alizarin, 97%, pure
Alizarin, 97%, pure
Thermo Scientific Chemicals
Alizarin, 97%, pure
CAS: 72-48-0 | C14H8O4 | 240.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA153691000
Price (MYR)
373.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
373.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAlizarin
Melting Point287.0°C to 289.0°C
Name Note97%
Boiling Point430.0°C
CAS72-48-0