Solketal, 97%
Solketal, 97%
Solketal, 97%
Thermo Scientific Chemicals
Solketal, 97%
CAS: 100-79-8 | C6H12O3 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA153655000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSolketal
Name Note97%
CAS100-79-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement