m-Toluoyl chloride, 99%
m-Toluoyl chloride, 99%
m-Toluoyl chloride, 99%
m-Toluoyl chloride, 99%
Thermo Scientific Chemicals
m-Toluoyl chloride, 99%
CAS: 1711-06-4 | C8H7ClO | 154.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA153411000
Price (MYR)
439.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
439.00
BOT
Add to cart
Specifications
Chemical Name or Materialm-Toluoyl chloride
Name Note99%
CAS1711-06-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger