o-Toluoyl chloride, 99%
o-Toluoyl chloride, 99%
o-Toluoyl chloride, 99%
o-Toluoyl chloride, 99%
Thermo Scientific Chemicals
o-Toluoyl chloride, 99%
CAS: 933-88-0 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA153371000
Price (MYR)
527.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
527.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Toluoyl chloride
Name Note99%
CAS933-88-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: