3,5-Dinitrobenzoic acid, 98+%
3,5-Dinitrobenzoic acid, 98+%
3,5-Dinitrobenzoic acid, 98+%
3,5-Dinitrobenzoic acid, 98+%
Thermo Scientific Chemicals
3,5-Dinitrobenzoic acid, 98+%
CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA153195000
Price (MYR)
666.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
666.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 5-Dinitrobenzoic acid
Name Note99%
CAS99-34-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning