Terephthaloyl Chloride, 99+%
Terephthaloyl Chloride, 99+%
Terephthaloyl Chloride, 99+%
Terephthaloyl Chloride, 99+%
Thermo Scientific Chemicals
Terephthaloyl Chloride, 99+%
CAS: 100-20-9 | C8H4Cl2O2 | 203.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA153102500
Price (MYR)
403.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTerephthaloyl chloride
Name Note99+%
CAS100-20-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.