3,3-Dimethyl-1-butene, 95%
3,3-Dimethyl-1-butene, 95%
3,3-Dimethyl-1-butene, 95%
3,3-Dimethyl-1-butene, 95%
Thermo Scientific Chemicals
3,3-Dimethyl-1-butene, 95%
CAS: 558-37-2 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA152720500
Price (MYR)
369.00
BOT
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
369.00
BOT
Add to cart
Specifications
Chemical Name or Material3,3-Dimethyl-1-butene
Name Note95%
CAS558-37-2
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger