3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
Thermo Scientific Chemicals
3,3-Dimethylpentane, 99%
CAS: 562-49-2 | C7H16 | 100.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA152660050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3, 3-Dimethylpentane
CAS562-49-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0