m-Anisic acid, 98%
m-Anisic acid, 98%
m-Anisic acid, 98%
Thermo Scientific Chemicals
m-Anisic acid, 98%
CAS: 586-38-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA152470250
Price (MYR)
323.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
323.00
BOT
Add to cart
Specifications
Chemical Name or Materialm-Anisic acid
CAS586-38-9
Health Hazard 1GHS Signal Word: Warning
Melting Point103.0°C to 108.0°C
ColorBeige-Yellow to White