p-Anisic acid, 98%
p-Anisic acid, 98%
p-Anisic acid, 98%
Thermo Scientific Chemicals
p-Anisic acid, 98%
CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number FSA152372500
Price (MYR)
483.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
483.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Anisic acid
Name Note98%
CAS100-09-4
Melting Point181.0°C to 186.0°C
ColorBeige-Gray to White