1-Benzylimidazole, 99%
1-Benzylimidazole, 99%
1-Benzylimidazole, 99%
1-Benzylimidazole, 99%
Thermo Scientific Chemicals
1-Benzylimidazole, 99%
CAS: 4238-71-5 | C10H10N2 | 158.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA152251000
Price (MYR)
2,428.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,428.00
EA
Add to cart
Specifications
Chemical Name or Material1-Benzylimidazole
CAS4238-71-5
CAS Min %98.5
CAS Max %100.0
Melting Point68.0°C to 71.0°C