2,2'-Biphenol, 99%
2,2'-Biphenol, 99%
2,2'-Biphenol, 99%
Thermo Scientific Chemicals
2,2'-Biphenol, 99%
CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA151991000
Price (MYR)
428.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
428.00
BOT
Add to cart
Specifications
Chemical Name or Material2,2′-Biphenol
Name Note99%
CAS1806-29-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning