2-Bromo-4'-methoxyacetophenone, 98%
2-Bromo-4'-methoxyacetophenone, 98%
2-Bromo-4'-methoxyacetophenone, 98%
2-Bromo-4'-methoxyacetophenone, 98%
Thermo Scientific Chemicals
2-Bromo-4'-methoxyacetophenone, 98%
CAS: 2632-13-5 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA151900250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-4'-methoxyacetophenone
CAS2632-13-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0