Hexanal, 96%, stabilized
Hexanal, 96%, stabilized
Hexanal, 96%, stabilized
Hexanal, 96%, stabilized
Thermo Scientific Chemicals
Hexanal, 96%, stabilized
CAS: 66-25-1 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA151855000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexanal
Acidity10mg KOH/g max.
CAS66-25-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.