Isophthalaldehyde, 99%
Isophthalaldehyde, 99%
Isophthalaldehyde, 99%
Isophthalaldehyde, 99%
Thermo Scientific Chemicals
Isophthalaldehyde, 99%
CAS: 626-19-7 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA151740250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsophthalaldehyde
Name Note99%
CAS626-19-7
Melting Point88°C to 90°C
ColorWhite to Yellow