1-Phenyldodecane, 97%
1-Phenyldodecane, 97%
1-Phenyldodecane, 97%
1-Phenyldodecane, 97%
Thermo Scientific Chemicals
1-Phenyldodecane, 97%
CAS: 123-01-3 | C18H30 | 246.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA151020100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Phenyldodecane
CAS123-01-3
CAS Min %96.0
CAS Max %100.0
Melting Point-7°C