Tributyrin, 97%
Tributyrin, 97%
Tributyrin, 97%
Thermo Scientific Chemicals
Tributyrin, 97%
Tributyrin, 97%, C15H26O6, CAS Number-60-01-5, butyrin, butyryl triglyceride, glycerol tributanoate, glycerol tributyrate, glyceroltributyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, tributin, tributyrin, tri-n-butyrin, 100mL, Liquid, 96% min. (GC), 02, 273, 305.0 deg.C to 310.0 deg.C | CAS: 60-01-5 | C15H26O6 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA150882500
Price (MYR)
532.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
532.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTributyrin
Name Note98%
CAS60-01-5
Melting Point-75°C
ColorClear colorless to light yellow