2,5-Dimethoxyaniline, 99%
2,5-Dimethoxyaniline, 99%
2,5-Dimethoxyaniline, 99%
2,5-Dimethoxyaniline, 99%
2,5-Dimethoxyaniline, 99%
CAS: 102-56-7 | C8H11NO2 | 153.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA150850500
Price (MYR)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 5-Dimethoxyaniline
Name Note99%
CAS102-56-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger