Acetanilide, 99+%
Acetanilide, 99+%
Acetanilide, 99+%
Thermo Scientific Chemicals
Acetanilide, 99+%
CAS: 103-84-4 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA150815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetanilide
CAS103-84-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0