Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Thermo Scientific Chemicals
Ethyl 4-aminobenzoate, 98%
CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA150785000
Price (MYR)
760.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
760.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl 4-aminobenzoate
CAS94-09-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a