D(+)-Mannose, 99+%
D(+)-Mannose, 99+%
D(+)-Mannose, 99+%
Thermo Scientific Chemicals
D(+)-Mannose, 99+%
CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA150601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Mannose
CAS3458-28-4
Melting Point133.0°C
ColorWhite
EINECS Number222-392-4