2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
Thermo Scientific Chemicals
2-Methyl-2,4-pentanediol, 99%
CAS: 107-41-5 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA150340010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Methyl-2, 4-pentanediol
CAS107-41-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0