2,6-Dimethylcyclohexanol, 99%, mixture of isomers
2,6-Dimethylcyclohexanol, 99%, mixture of isomers
2,6-Dimethylcyclohexanol, 99%, mixture of isomers
2,6-Dimethylcyclohexanol, 99%, mixture of isomers
Thermo Scientific Chemicals
2,6-Dimethylcyclohexanol, 99%, mixture of isomers
CAS: 5337-72-4 | C8H16O | 128.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA149810500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Dimethylcyclohexanol
Name Note99%
CAS5337-72-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning