Di-n-butyl ether, 99+%, extra pure
Di-n-butyl ether, 99+%, extra pure
Di-n-butyl ether, 99+%, extra pure
Di-n-butyl ether, 99+%, extra pure
Thermo Scientific Chemicals
Di-n-butyl ether, 99+%, extra pure
CAS: 142-96-1 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA149690010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-butyl ether
CAS142-96-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN