DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
Thermo Scientific Chemicals
DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol
CAS: 78-96-6 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass bottle
Catalog number FSA149650025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-1-Amino-2-propanol
Name Note99+%
CAS78-96-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement