Acrylonitrile, 99+%
Acrylonitrile, 99+%
Acrylonitrile, 99+%
Thermo Scientific Chemicals
Acrylonitrile, 99+%
CAS: 107-13-1 | C3H3N | 53.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA149631000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylonitrile
CAS107-13-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0