(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
(±)-Propylene oxide, 99%, pure
Thermo Scientific Chemicals
(±)-Propylene oxide, 99%, pure
(+/-)-Propylene oxide, 99%, C3H6O, CAS Number-75-56-9, 2,3-epoxypropane, epoxypropane, methyloxirane, propylene oxide, oxirane, methyl, propyleneoxide, 1,2-propylene oxide, propene oxide, propylene epoxide, 1,2-epoxypropane, 1L, 17, 6, 34.0 deg.C, CHEBI:38685, Undesignated, 0.8300g/mL, 200-879-2 | CAS: 75-56-9 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA149620010
Price (MYR)
359.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
359.00
BOT
Add to cart
Specifications
Chemical Name or Material(±)-Propylene oxide
Name Note99%
CAS75-56-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.
May cause genetic defects.