Tetraethylene glycol, 99.5%
Tetraethylene glycol, 99.5%
Tetraethylene glycol, 99.5%
Tetraethylene glycol, 99.5%
Thermo Scientific Chemicals
Tetraethylene glycol, 99.5%
CAS: 112-60-7 | C8H18O5 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA149590025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraethylene glycol
CAS112-60-7
Melting Point-4.1°C
ColorYellow
Density1.1200g/mL