Ethanolamine, 99%
Ethanolamine, 99%
Ethanolamine, 99%
Thermo Scientific Chemicals
Ethanolamine, 99%
CAS: 141-43-5 | C2H7NO | 61.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
25 L
Catalog number FSA149582500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthanolamine
Name Note99%
CAS141-43-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement