Valeric Acid, 99%
Valeric Acid, 99%
Valeric Acid, 99%
Thermo Scientific Chemicals
Valeric Acid, 99%
CAS: 109-52-4 | C5H10O2 | 102.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA149571000
Price (MYR)
394.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
EA
Add to cart
Specifications
Chemical Name or MaterialValeric acid
CAS109-52-4
Melting Point-20.0°C to +18.0°C
Boiling Point185.0°C
Health Hazard 1Danger