Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Thermo Scientific Chemicals
Phenol, 99%, extra pure
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
50 g
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA149340051
Price (MYR)
1,917.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,917.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenol
Name Note99%
CAS108-95-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated