2-Picoline, 98%
2-Picoline, 98%
2-Picoline, 98%
Thermo Scientific Chemicals
2-Picoline, 98%
CAS: 109-06-8 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA149330250
Price (MYR)
228.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
228.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Picoline
Name Note98%
CAS109-06-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger