Isopropanol, 99.5+%, pure
Isopropanol, 99.5+%, pure
Isopropanol, 99.5+%, pure
Isopropanol, 99.5+%, pure
Thermo Scientific Chemicals
Isopropanol, 99.5+%, pure
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
View all
Plastic Drum
Catalog number FSA149320050
Price (MYR)
1,164.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,164.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Name Note99.8+%
CAS67-63-0
Health Hazard 1Danger