Propylamine, 98%, pure, Thermo Scientific Chemicals
Propylamine, 98%, pure, Thermo Scientific Chemicals
Propylamine, 98%, pure, Thermo Scientific Chemicals
Propylamine, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propylamine, 98%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA149310010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPropylamine
Name Note98%
CAS107-10-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.