o-Anisaldehyde, 98%
o-Anisaldehyde, 98%
o-Anisaldehyde, 98%
Thermo Scientific Chemicals
o-Anisaldehyde, 98%
CAS: 135-02-4 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA149221000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialo-Anisaldehyde
Name Note98%
CAS135-02-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning