4-Methoxybenzenethiol, 98%
4-Methoxybenzenethiol, 98%
4-Methoxybenzenethiol, 98%
4-Methoxybenzenethiol, 98%
Thermo Scientific Chemicals
4-Methoxybenzenethiol, 98%
CAS: 696-63-9 | C7H8OS | 140.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA149150250
Price (MYR)
2,440.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,440.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxybenzenethiol
Name Note98%
CAS696-63-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning