Saccharin, 98+%
Saccharin, 98+%
Saccharin, 98+%
Thermo Scientific Chemicals
Saccharin, 98+%
CAS: 81-07-2 | C7H5NO3S | 183.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA149001000
Price (MYR)
313.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
313.00
EA
Add to cart
Specifications
Chemical Name or MaterialSaccharin
CAS81-07-2
Melting Point226.0°C to 230.0°C
ColorWhite
EINECS Number201-321-