Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%
Thermo Scientific Chemicals
Thiamine hydrochloride, 98.5-101.5%
Thiamine hydrochloride, 98.5-101.5%, C12H17ClN4OS.HCl, CAS Number-67-03-8, aneurine hydrochloride, berin, thiamin chloride, thiamine chloride hydrochloride, thiamine dichloride, thiamine hcl, thiamine hydrochloride, thiaminium chloride, trophite, vitamin b1 hydrochloride, 1kg, 98.5 to 101.5% | CAS: 67-03-8 | C12H17ClN4OS·HCl | 337.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA148991000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThiamine hydrochloride
Name Note98.5-101.5%
CAS67-03-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: